Learn More
CD11a/CD18 Antibody (hu1124 (Efalizumab)), Biotin, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
CD11a/CD18 Monoclonal antibody specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization
Spezifikation
Spezifikation
| Antigen | CD11a/CD18 |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Neutralization |
| Klassifikation | Monoclonal |
| Klon | hu1124 (Efalizumab) |
| Konjugat | Biotin |
| Verdünnung | Flow Cytometry, Immunohistochemistry, Block/Neutralize |
| Zusammensetzung | PBS |
| Gen-Alias | CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 |
| Wirtsspezies | Human |
| Immunogen | Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes. |
| Mehr anzeigen |