Learn More
Thermo Scientific™ Acetylcholine iodide, 98%
Artikelnummer. 15908046
Ansicht ändern
Click to view available options
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15908046 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15908046 Lieferant Thermo Scientific™ Lieferanten-Nr. A11694.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Spezifikation
CAS 2260-50-6
Schmelzpunkt 161-165°
Summenformel C{7}H{1}{6}INO{2}
Strukturformel CH{3}CO{2}CH{2}CH{2}N(CH{3}){3}I
MDL-Nummer MFCD00011815
Menge 5 g
Synonym (2-Acetoxyethyl)trimethylammonium iodide;2-Acetoxy-N,N,N-trimethylethanium iodide
Formelmasse 273.11
Reinheit (%) 98%
Physikalische Form Solid