missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholinesterase/ACHE Antibody, Novus Biologicals™

Artikelnummer. 18074014 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
Product Code. Menge unitSize
18074014 100 μL 100 Mikroliter
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18074014

missing translation for 'mfr': Novus Biologicals NBP131329

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylcholinesterase/ACHE
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 1.0mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000
Zusammensetzung PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gen-Zugriffsnummer P22303
Gen-Alias acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group
Gensymbole ACHE
Wirtsspezies Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human AChE. The exact sequence is proprietary.
Molekulargewicht des Antigens 68 kDa
Reinigungsverfahren Affinity Purified
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primär oder sekundär Primary
Gen-ID (Entrez) 43
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.