Learn More
Acetylcholinesterase/ACHE Antibody, PE, Novus Biologicals™
Goat Polyclonal Antibody
Marke: Novus Biologicals NB600-911PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Acetylcholinesterase/ACHE Polyclonal antibody specifically detects Acetylcholinesterase/ACHE in Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Spezifikation
| Acetylcholinesterase/ACHE | |
| Polyclonal | |
| PBS | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Bovine | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| PE | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Acetylcholinesterase/ACHE [Bovine Erythrocytes] (Uniprot: P23795) | |
| 0.1 mL | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 43 | |
| Store at 4°C in the dark. |