Learn More
Invitrogen™ ACTA1/ACTA2/ACTC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5100907
Beschreibung
Antibody detects endogenous levels of total ACTA1/A2/C1.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Spezifikation
| ACTA1/ACTA2/ACTC1 | |
| Polyclonal | |
| Unconjugated | |
| Acta1 | |
| 0610041G09Rik; AA959943; AAT6; Acta; Acta1; Acta2; Acta-2; ACTC; ACTC1; Actc-1; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; Acts; ACTSA; Actsk-1; Actvs; alpha-actin cardiac; alpha-actin-1; Alpha-actin-2; alphac-actin; alpha-cardiac actin; alphaSMA; ASD5; ASMA; a-SMA; cell growth-inhibiting gene 46 protein; CFTD; CFTD1; CFTDM; CMD1R; CMH11; GIG46; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; SMalphaA; smooth muscle alpha-actin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11459, 11464, 11475, 29275, 29437, 58, 59, 70, 81633 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62736, P62737, P62738, P68032, P68033, P68035, P68133, P68134, P68136 | |
| Acta1, ACTA2, Actc1 | |
| A synthesized peptide derived from human ACTA1(Accession P68133), corresponding to amino acid residues M1-P34. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.