Learn More
ERLAB BE Activated Carbon Filter
Artikelnummer. 30484798
Click to view available options
Zur Verwendung mit (Anwendung):
Säuren und Lösungsmittel
Packungsgröße:
1 Stück
Artikelnummer. 30484798
Marke: ERLAB NORT000478/BE
om dit product te kopen Registreer vandaag om een webaccount aan te maken
BE - Extraction filter targets acids and solvent
  • Activated Carbon - BE - Extraction filter is tailored for capturing acidic vapors and organic solvents
  • Ideal for laboratories handling corrosive and volatile chemical substances
  • Dual-action filtration prolongs filter life and increases safety
  • Prevents harmful emissions from being released into the workspace
  • Ensures compliance with health and safety regulations for air quality in laboratories
  • Recommended for multi-substance handling where both acids and solvents are present
Spezifikation
Zur Verwendung mit (Anwendung) Säuren und Lösungsmittel