Learn More
Upchurch Scientific Adapter thread to barbed for 1/16 inch to 3/32inch (1.55-2.40mm) I.D. tubing PEEK(TM) to 1/16inc
Marke:  Upchurch Scientific 02023-82
Adapter thread to barbed for 1/16 inch to 3/32inch (1.55-2.40mm) I.D. tubing PEEK(TM) to 1/16inc
Artikelnummer. 13020440
  • 29.05 € / 1 Stück
Nur noch null auf Lager
Verfügbar ab: 20-02-2026