Learn More
Thermo Scientific Chemicals Adenosin, 99 %
Artikelnummer. p-7080986
Click to view available options
Menge:
10 g
50 g
250 g
Packungsgröße:
10 Gramm
250 Gramm
50 Gramm
Artikelnummer. 11439692
Marke: Thermo Scientific Chemicals A10781.09
Bulk oder benutzerdefiniertes Format anfordern
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Edge
CAS: 58-61-7 | C10H13N5O4 | 267.25 g/mol
Adenosine serves as a nucleotide. It also plays an important role in the regulation of blood flow to various organs through vasodilation. It is actively involved in biochemical processes, such as energy transfer as adenosine triphosphate and adenosine diphosphate. Further, it is used in medication specifically as an antiarrhythmic agent to treat a number of supraventricular tachycardia. It is used to promote thickening of hair for people with thinning hair.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Löslichkeit
Löslich in Wasser, Ammoniumhydroxid und Dimethylsulfoxid. Nicht löslich in Ethanol.
Hinweise
Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 58-61-7
Summenformel C10H13N5O4
Molekulargewicht (g/mol) 267.25
MDL-Nummer MFCD00005752
InChI-Schlüssel OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC-Name (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Spezifikation
Chemischer Name oder Material Adenosine
Schmelzpunkt 234°C to 237°C
Prozentgehaltsbereich 99%
Geruch Geruchsneutral
Menge 10 g
Beilstein 93029
Merck Index 14,153
Löslichkeitsinformationen Soluble in water,ammonium hydroxide and dimethyl sulfoxide. Insoluble in ethanol.
Optische Drehung −70° (c=2 in 5% NaOH)
Formelmasse 267.25
Reinheit (%) 99%