missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 5/6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538844
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a member of the membrane-bound adenylyl cyclase enzymes. Adenylyl cyclases mediate G protein-coupled receptor signaling through the synthesis of the second messenger cAMP. Activity of the encoded protein is stimulated by the Gs alpha subunit of G protein-coupled receptors and is inhibited by protein kinase A, calcium and Gi alpha subunits. Single nucleotide polymorphisms in this gene may be associated with low birth weight and type 2 diabetes. Alternatively spliced transcript variants that encode different isoforms have been observed for this gene. [provided by RefSeq, Dec 2010].
Spezifikation
| Adenylate Cyclase 5/6 | |
| Polyclonal | |
| Unconjugated | |
| ADCY5 | |
| ADCY5, ADCY6 | |
| A synthetic peptide derived from the internal region of human ADCY5/6 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43306, O95622, P84309, Q01341, Q03343, Q04400 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 111, 112, 11512, 224129, 25289, 64532 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur