missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Adiponectin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Artikelnummer. 17895153
Change view
Click to view available options
Menge:
2 x 500 μg
25 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
25 Mikrogramm
50 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
17895153 50 μg 50 Mikrogramm
17894863 25 μg 25 Mikrogramm
17815673 2 x 500 μg 1 Milligramm
30054755 500 μg 500 Mikrogramm
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17895153

Marke: PeproTech 500P239BT50UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: RGHHHHHHHH ETTTQGPGVL LPLPKGACTG WMAGIPGHPG HNGAPGRDGR DGTPGEKGEK GDPGLIGPKG DIGETGVPGA EGPRGFPGIQ GRKGEPGEGA YVYRSAFSVG LETYVTIPNM PIRFTKIFYN QQNHYDGSTG KFHCNIPGLY YFAYHITVYM KDVKVSLFKK DKAMLFTYDQ YQENNVDQAS GSVLLHLEVG DQVWLQVYGE GERNGLYADN DNDSTFTGFL LYHDTN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Adiponectin. Anti-Human Adiponectin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Adiponectin by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Adiponectin (500-P239) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human Adiponectin. Western Blot: To detect Human Adiponectin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Adiponectin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P239BT-1MG will be provided as 2 x 500 μg

Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Adiponectin
Anwendungen ELISA, Western Blot
Klassifikation Polyclonal
Konzentration 0.1-1.0 mg/mL
Konjugat Biotin
Zusammensetzung PBS with no preservative
Gen ADIPOQ
Gen-Zugriffsnummer Q15848
Gen-Alias 30 kDa adipocyte complement-related protein; 30kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28
Gensymbole ADIPOQ
Wirtsspezies Rabbit
Immunogen Hi-5 Insect cells-derived Recombinant Human Adiponectin.
Reinigungsverfahren Antigen affinity chromatography
Menge 50 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 9370
Zielspezies Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Lyophilized
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.