missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adrenomedullin/ADM Antibody (HTA 91a/G2), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB100-2632-0.025ml
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Adrenomedullin/ADM Monoclonal antibody specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Spezifikation
| Adrenomedullin/ADM | |
| Monoclonal | |
| 2 mg/mL | |
| Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| adrenomedullin, AMpreproadrenomedullin | |
| Chemically synthesized full length adrenomedullin | |
| 0.025 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience | |
| 133 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HTA 91a/G2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion