Learn More
Invitrogen™ ADRM1 Recombinant Rabbit Monoclonal Antibody (24GB4700)
Beschreibung
ADRM1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is an integral plasma membrane protein which promotes cell adhesion. The encoded protein is thought to undergo O-linked glycosylation. Expression of this gene has been shown to be induced by gamma interferon in some cancer cells. Two transcript variants encoding the same protein have been found for this gene.
Spezifikation
Spezifikation
| Antigen | ADRM1 |
| Anwendungen | Flow Cytometry, Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB4700 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | ADRM1 |
| Gen-Zugriffsnummer | Q16186 |
| Gen-Alias | 110 kDa cell membrane glycoprotein; 1110063P18Rik; 2510006J17Rik; AA408205; adhesion regulating molecule 1; adhesion-regulating molecule 1; ADRM1; Arm1; ARM-1; AU043535; cell membrane glycoprotein, 110000M(r) (surface antigen); glycoprotein 110; GP110; hRpn13; M(r) 110,000 surface antigen; MGC29536; proteasomal ubiquitin receptor ADRM1; Proteasome regulatory particle non-ATPase 13; Rpn13; rpn13 homolog |
| Gensymbole | ADRM1 |
| Mehr anzeigen |