missing translation for 'onlineSavingsMsg'
Learn More

ADRP/Perilipin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Artikelnummer. 16824414 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16824414 150 μL 150 Mikroliter
16834414 20 μL 20 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16824414

Marke: Proteintech 152941AP150UL

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Anti-ADRP antibody is a reliable and sensitive marker for lipid droplets. Enhanced expression of ADRP is linked to diseases with abnormal lipid storage, including hepatic steatosis, atherosclerosis and diabetes. Immunohistochemistry of ADRP may facilitate

Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.

Spezifikation

Antigen ADRP/Perilipin 2
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 0.35 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen PLIN2
Gen-Zugriffsnummer P43883, Q99541
Gen-Alias ADFP, Adipophilin, ADRP, Perilipin 2, PLIN2
Gensymbole Plin2
Wirtsspezies Rabbit
Immunogen ADRP/Perilipin 2 Fusion Protein Ag7539
Reinigungsverfahren Antigen Affinity Chromatography
Menge 150 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11520, 123, 298199
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.