Beschreibung
The AGO2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human AGO2. This antibody recognizes human, mouse, rat antigen. The AGO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
| Antigen | AGO2 |
| Anwendungen | Western Blot, Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | 1G2H12 |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Menge | 20 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 27161 |
| Zielspezies | Human, Mouse, Rat |
| Mehr anzeigen |