Learn More
Invitrogen™ AGPS Recombinant Rabbit Monoclonal Antibody (24GB6810)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA556957
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30276062
  • 514.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
AGPS Recombinant Monoclonal Antibody for Western Blot
This gene is a member of the FAD-binding oxidoreductase/transferase type 4 family. It encodes a protein that catalyzes the second step of ether lipid biosynthesis in which acyl-dihydroxyacetonephosphate (DHAP) is converted to alkyl-DHAP by the addition of a long chain alcohol and the removal of a long-chain acid anion. The protein is localized to the inner aspect of the peroxisomal membrane and requires FAD as a cofactor. Mutations in this gene have been associated with rhizomelic chondrodysplasia punctata, type 3 and Zellweger syndrome.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
AGPS
Recombinant Monoclonal
Unconjugated
AGPS
5832437L22; 9930035G10Rik; AAG5; Adaps; Adap-s; ADAS; Adhaps; ADPS; Ads; Aging-associated gene 5 protein; aging-associated protein 5; AGPS; ALDHPSY; alkyl-DHAP synthase; alkyldihydroxyacetone phosphate synthase; alkyl-dihydroxyacetonephosphate synthase; alkyldihydroxyacetonephosphate synthase, peroxisomal; alkylglycerone phosphate synthase; alkylglycerone phosphate synthase-like; alkylglycerone-phosphate synthase; AW123847; bs2; RCDP3
Rabbit
Affinity chromatography
RUO
84114, 8540
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6810
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00116, Q9EQR2
AGPS
Recombinant protein of human Alkyl-DHAP synthase (aa 71-182).
100 μL
Primary
Human, Rat