missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHCY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10757-2-AP-150UL
This item is not returnable.
View return policy
Description
S-adenosylhomocysteine hydrolase (AHCY) catalyzes the reversible hydrolysis of S-adenosylhomocysteine (AdoHcy) to adenosine (Ado) and L-homocysteine (Hcy). Thus, it regulates the intracellular S-adenosylhomocysteine (SAH) concentration thought to be important for transmethylation reactions. Deficiency in this protein is one of the different causes of hypermethioninemia. S-adenosylhomocysteine hydrolase belongs to the adenosylhomocysteinase family.Specifications
| AHCY | |
| Polyclonal | |
| Unconjugated | |
| AHCY | |
| Adenosylhomocysteinase, AdoHcyase, AHCY, S adenosylhomocysteine hydrolase, SAHH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 191, 269378, 29443 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10760, P23526, P50247 | |
| AHCY | |
| AHCY Fusion Protein Ag1208 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction