Beschreibung
AIF-1/Iba1 Polyclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Spezifikation
Spezifikation
| Antigen | AIF-1/Iba1 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA |
| Klassifikation | Polyclonal |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1 - 3 ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:16000 |
| Zusammensetzung | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gen-Zugriffsnummer | NP_116573.1 |
| Gen-Alias | AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 |
| Gensymbole | AIF1 |
| Mehr anzeigen |