missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIMP2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-81885FR
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
AIMP2 Polyclonal antibody specifically detects AIMP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
| AIMP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| aminoacyl tRNA synthase complex-interacting multifunctional protein 2, aminoacyl tRNA synthetase complex-interacting multifunctional protein 2, ARS-interacting multi-functional protein 2, JTV1JTV-1, Multisynthase complex auxiliary component p38, multisynthetase complex auxiliary component p38, p38, PRO0992, Protein JTV-1 | |
| AIMP2 antibody was raised against a 16 amino acid peptide near the center of human AIMP2. The immunogen is located within amino acids 150 - 200 of AIMP2. Amino Acid Squence: FGEQNKKQPRQDYQL | |
| 0.1 mL | |
| Apoptosis | |
| 7965 | |
| Store at 4°C in the dark. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur