missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-32560
This item is not returnable.
View return policy
Description
AK7 Polyclonal specifically detects AK7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| AK7 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| AK7 | |
| Recombinant protein encompassing a sequence within the center region of human Adenylate kinase 7. The exact sequence is proprietary. | |
| 100 μL | |
| Lipid and Metabolism | |
| 122481 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| adenylate kinase 7, EC 2.7.4.3, FLJ32864, putative adenylate kinase 7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction