Learn More
Albumin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-32458
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Albumin Polyclonal antibody specifically detects Albumin in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Spezifikation
| Albumin | |
| Polyclonal | |
| Rabbit | |
| 69 kDa | |
| RUO |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| ALB | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Albumin. The exact sequence is proprietary. | |
| 0.1 mL | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |