missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldicarb Sulfone, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Aldicarb-sulfone |
| CAS | 1646-88-4 |
| Summenformel | C7 H14 N2 O4 S |
| Menge | 50 mg |
| Synonym | Propionaldehyde, 2-methyl-2-(methylsulfonyl)-, O-(methylcarbamoyl)oxime (7CI,8CI), Carbamic acid, methyl-, O-[[2-methyl-2-(methylsulfonyl)propylidene]amino] deriv. (8CI), 2-Methyl-2-(methylsulfonyl)propionaldehyde O-(methylcarbamoyl)oxime, Aldicarb sulfone, Aldoxycarb, Standak, Sulfocarb, Temik sulfone, UC 21865, UCES, Yangdimiewei |
| InChI-Formel | InChI=1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10) |
| SMILES | CNC(=O)ON=CC(C)(C)S(=O)(=O)C |
| IUPAC-Name | [(2-methyl-2-methylsulfonylpropylidene)amino] N-methylcarbamate |
| Molekulargewicht (g/mol) | 222.26 |
| Formelmasse | 222.0674 |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?