missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (H17E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
216.00 € - 527.00 €
Spezifikation
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Klon | H17E2 |
| Konzentration | 1.0 mg/mL |
| Verdünnung | Flow Cytometry 10 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Anwendungen | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18214305
|
Novus Biologicals
NB600-750-0.025MG |
0.025 mg |
216.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18007583
|
Novus Biologicals
NB600-750 |
0.1 mg |
527.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Spezifikation
| Alkaline Phosphatase/ALPP | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| ALPP | |
| IgG1 | |
| Protein G purified | |
| This recognises human placental alkaline phosphate. The has been widely used in immunolocalisation studies of germ cell tumours and may also be used in ELISA based assays for the measurement of soluble PLAP. |
| H17E2 | |
| Flow Cytometry 10 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 250 | |
| Placental membrane (Human). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts