missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Smooth Muscle Actin Antibody (1A4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB1420F
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
alpha-Smooth Muscle Actin Monoclonal antibody specifically detects alpha-Smooth Muscle Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, Dual RNAscope ISH-IHC
Spezifikation
| alpha-Smooth Muscle Actin | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 1A4 | |
| Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC | |
| AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin | |
| Peptide corresponding to residues EEEDSTALVC of the N-term of human alpha-smooth muscle actin (1) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 59 | |
| Store at 4C in the dark. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur