missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (2A7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
254.00 € - 623.00 €
Spezifikation
| Antigen | alpha-Synuclein |
|---|---|
| Klon | 2A7 |
| Konzentration | 1 mg/ml |
| Verdünnung | Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 |
| Anwendungen | Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18074866
|
Novus Biologicals
NBP1-05194-0.025ML |
0.025 mL |
254.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18017054
|
Novus Biologicals
NBP1-05194 |
0.1 mL |
623.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Spezifikation
| alpha-Synuclein | |
| 1 mg/ml | |
| Immunohistochemistry (Free Floating), Immunofluorescence, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Pig, Human, Bovine, Rat, Mouse | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| SNCA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2A7 | |
| Western Blot 1:1000, Simple Western 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| P37840 | |
| 6622 | |
| IgG1 | |
| Immunogen affinity purified | |
| 15 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts