Learn More
Aluminum, Al/Mg, AA 6063, A96063, ARMI∣MBH
Click to view available options
Menge:
50 g
Packungsgröße:
50 Gramm
Beschreibung
Aluminum, Al/Mg, AA 6063, A96063
Spezifikation
Spezifikation
| Chemischer Name oder Material | Boron, Chromium, Copper, Gallium, Iron, Lead, Magnesium, Manganese, Nickel, Silicon, Tin, Titanium, Vanadium, Ytterbium, Zinc, Zirconium |
| Menge | 50 g |
| Gewicht | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Zirconium: 0.0008%, Lead: 0.001%, Boron: 0.0014%, Tin: 0.0014%, Nickel: 0.0066%, Chromium: 0.011%, Zinc: 0.0122%, Titanium: 0.013%, Vanadium: 0.0131%, Gallium: 0.0139%, Copper: 0.023%, Manganese: 0.039%, Iron: 0.203%, Silicon: 0.47%, Magnesium: 0.56%, Ytterbium <0.001 ppm |