missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AM 2394, Tocris Bioscience™
Potent glucokinase activator; orally bioavailable
Spezifikation
| Chemischer Name oder Material | AM 2394 |
|---|---|
| Physikalische Form | Solid |
| Produkttyp | Bioactive Small Molecule |
| Löslichkeit | Soluble to 100mM in DMSO and to 10mM in ethanol with gentle warming |
| Synonym | N-[6'-(2-Hydroxy-2-methylpropoxy)-4-[(5-methyl-3-pyridinyl)oxy][3,3'-bipyridin]-6-yl]-N'-methylurea |
Beschreibung
Potent glucokinase activator (EC50 = 60 nM). Orally bioavailable. Reduces plasma glucose levels in an oral glucose tolerance test in ob/ob mice.Spezifikation
| AM 2394 | |
| Bioactive Small Molecule | |
| N-[6'-(2-Hydroxy-2-methylpropoxy)-4-[(5-methyl-3-pyridinyl)oxy][3,3'-bipyridin]-6-yl]-N'-methylurea |
| Solid | |
| Soluble to 100mM in DMSO and to 10mM in ethanol with gentle warming | |
| 1442684-77-6 |
Sicherheit und Handhabung
Recommended Storage : Store at -20°C
For Research Use Only.
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.