missing translation for 'onlineSavingsMsg'
Learn More

AMG 487, MedChemExpress

Artikelnummer. 16496448
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
10 Milligramm
100 Milligramm
5 Milligramm
50 Milligramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16496448 10 mg 10 Milligramm
16469037 5 mg 5 Milligramm
16446736 100 mg 100 Milligramm
16432557 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. View return policy
Product Code. 16496448 Supplier Medchem Express Supplier No. HY15319

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively.

AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively.

Specifications

Chemischer Name oder Material AMG 487
CAS 473719-41-4
Farbe Yellow
Hinweise zur Reinheitsqualität Research
Summenformel C32H28F3N5O4
Menge 10 mg
Löslichkeitsinformationen DMSO : ≥ 41 mg/mL (67.93 mM)
SMILES O=C(N([C@@H](C1=NC2=NC=CC=C2C(N1C3=CC=C(OCC)C=C3)=O)C)CC4=CC=CN=C4)CC5=CC=C(OC(F)(F)F)C=C5
Molekulargewicht (g/mol) 603.59
Formelmasse 603.59
Reinheit (%) 99.17%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.