Learn More
Melford Ammonium Chloride
Click to view available options
Menge:
500 g
1 kg
Packungsgröße:
1 Kilogramm
500 Gramm
Beschreibung
Ammonium chloride, an inorganic compound, is a white crystalline salt, highly soluble in water.
Ammonium chloride, an inorganic compound, is a white crystalline salt, highly soluble in water.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Ammonium Chloride |
| CAS | 12125-02-9 |
| Menge | 1 kg |
| Summenformel | NH4Cl |
| Güte | ACS |