missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate Tetrahydrate, Analytical Grade, Penta Chemicals

Artikelnummer. 30020491
Change view
Click to view available options
Menge:
250 g
500 g
Packungsgröße:
250 Gramm
500 Gramm
Artikelnummer. Menge unitSize
30020491 500 g 500 Gramm
30020622 250 g 250 Gramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30020491

Marke: PENTA CHEMICALS 2175030500

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Chemische Identifikatoren

CAS 12054-85-2
Molekulargewicht (g/mol) 1235.86

Spezifikation

Chemischer Name oder Material Ammonium Molybdate Tetrahydrate
Physikalische Form Crystalline Powder, Needles or Crystals
Menge 500 g
Reinheit (%) ≥99%
Güte Analytical
Verpackung Plastic Jar
compliance-icons
Produktidentifikator
  • Ammonium molybdate tetrahydrate
Signalwort
  • Achtung
Gefahrenkategorie
  • Schwere Augenschädigung/Augenreizung Kategorie 2
  • Hautreizende/-ätzende Wirkung Kategorie 2
  • Spezifische Zielorgan-Toxizität Kategorie 3
Gefahrenhinweis
  • H315-Verursacht Hautreizungen.
  • H319-Verursacht schwere Augenreizung.
  • H335-Kann die Atemwege reizen.
Sicherheitshinweis
  • P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P305+P351+P338-BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.
Ergänzende Informationen
  • MIXTURE LIST-Enthält: Ammonium molybdate tetrahydrate
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.