Learn More
TFA Dostmann Analog Maxima/Minima Thermometer
Beschreibung
With the Maxima-Minima-Thermometer you always have three values in view at the same time: the current, the highest and the lowest temperatures. You can determine the observation period yourself. Just press the pushbutton to reset the display to the current temperature. This device is weatherproof and not only ideal for gardening and agriculture, but also for temperature monitoring of living rooms, greenhouses, wine cellars, storage rooms or freezers. The capillary is filled with a mercury-free, patented fluid. This product is entirely manufactured in Germany.
- Weatherproof
- Made in Germany
- Display of current, highest and lowest temperature
- With patented, mercury-free liquid
- Suitable for indoor and outdoor use
Spezifikation
Spezifikation
| Material | Kunststoff |