missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Angiotensin (1-7)

Artikelnummer. 16415148
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
16415148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16415148 Supplier MedChemExpress Supplier No. HY12403

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress Angiotensin 1-7 (Ang-(1-7)) is an endogenous heptapeptide from the renin-angiotensin system (RAS) with a cardioprotective role due to its anti-inflammatory and anti-fibrotic activities in cardiac cells.

Angiotensin 1-7 inhibits purified canine ACE activity (IC50=0.65 μM). Angiotensin 1-7 acts as a local synergistic modulator of kinin-induced vasodilation by inhibiting ACE and releasing nitric oxide. Angiotensin 1-7 blocks Ang II-induced smooth muscle cell proliferation and hypertrophy and shows antiangiogenic and growth-inhibitory effects on the endothelium. Angiotensin 1-7 shows anti-inflammatory activity .

Specifications

Reinheit 98.04%
CAS 51833-78-4
Inhalt und Lagerung Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Zur Verwendung mit (Anwendung) COVID-19-anti-virus
Molekulargewicht 899.0 g/mol
Produkttyp Peptides
Menge 10 mg
Versandbedingung Room temperature
Löslichkeit H2O : ≥ 30.2 mg/mL (33.59 mM)
Synonym Ang-(1-7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.