missing translation for 'onlineSavingsMsg'
Learn More

Anisole-2,4,6-d3, CDN

Product Code. 30314746
Change view
Click to view available options
Menge:
0.5 g
1 g
Unit Size:
0.50 Gramm
1 Gramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30314746 1 g 1 Gramm
30328563 0.5 g 0.50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314746 Supplier CDN Supplier No. CDND11991G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Entdecken Sie über 4000 hochwertige deuterierte Verbindungen, die direkt ab Lager versandfertig sind und somit schnell geliefert werden. Wir bieten viele einzigartige APIs an, darunter pharmazeutische Wirkstoffe, Nitrosamine, Aminosäuren, Steroide, Gase, Pestizide und Fettsäuren.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Anisole-2,4,6-D3
CAS 2567-25-1
Summenformel C7 D3 H5 O
Menge 1 g
Synonym Benzene-1,3,5-d3, 2-methoxy- (9 CI, ACI), 2-Methoxybenzene-1,3,5-d3 (ACI), Anisole-2,4,6-d3 (6 CI, 7 CI, 8 CI), 1,3,5-Trideuterio-2-methoxybenzene, 2,4,6-Trideuterioanisole, 2,4,6-Trideuteroanisole, Methoxybenzene-2,4,6-d3, Anisole-2,4,6-D3
InChI-Formel InChI=1 S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6 H,1H3/i2D,5 D,6 D
SMILES [2 H]c1cc([2 H])c(OC)c([2 H])c1
IUPAC-Name 1,3,5-Trideuterio-2-methoxybenzol
Molekulargewicht (g/mol) 111.1563
Formelmasse 111.0763 g/mol
Reinheit (%) 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.