Learn More
antizyme inhibitor 1, Mouse, Clone: 8B9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AZIN1.
Marke: Abnova H00051582-M01.100ug
Beschreibung
Ornithine decarboxylase (ODC) catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. Ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein encoded by this gene is highly similar to ODC. It binds to ODC antizyme and stabilizes ODC, thus inhibiting antizyme-mediated ODC degradation. Two alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: HKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAESpezifikation
| antizyme inhibitor 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_015878 | |
| AZIN1 | |
| AZIN1 (NP_056962, 339 a.a. ∼ 447 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 8B9 | |
| Mouse monoclonal antibody raised against a partial recombinant AZIN1. | |
| AZIN1 | |
| MGC3832/MGC691/OAZI/OAZIN/ODC1L | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 51582 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.