Learn More
B-Raf Mouse, Unlabeled, Clone: 13, BD
Mouse Monoclonal Antibody
321.00 € - 459.19 €
Spezifikation
| Antigen | B-Raf |
|---|---|
| Klon | 13 |
| Konzentration | 250μg/mL |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
Beschreibung
The Raf kinase family, A-Raf, B-Raf, and C-Raf, act as downstream effectors for Ras. These ser/thr kinases have homologous kinase domains referred to as conserved regions (CR1, CR2, and CR3). Rafs have a critical role in the response to many growth factors including: EGF, PDGF, insulin, IL-2, IL-3, CSF-1, and GM-CSF. Raf can directly interact with Ras-GTP and subsequently become activated. This leads to Raf activation of the MAP kinase pathway by phosphorylating and activating MEK, which then phosphorylates and activates MAP kinaes (ERK).
A-Raf is a proto-onocogene that is highly expressed in urogenital and kidney tissues while B-Raf is expressed at high levels in cerebrum and testes. C-Raf appears to be uniformly and ubiquitously expressed. Cell transformation by A-Raf has been shown to be independent of p21ras, suggesting the possibility of an independent signal transduction pathway. B-Raf is expressed as multiple isoforms that range from 69-72kDa and 79-99kDa. In hematopoietic cells, B-Raf is activated by EPO, GM-CSF, and SCF. Thus, the Raf kinases are important Ras effectors involved in growth factor signaling in a wide variety of cells and tissues.
Western Blotting
Spezifikation
| B-Raf | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 13 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Human B-Raf aa. 285-406 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.