Learn More
CD123 Mouse anti-Human, PE, Clone: 9F5, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561050
Beschreibung
The 9F5 monoclonal antibody specifically binds to CD123. CD123 is the 70 kDa IL-3 receptor α chain (IL-3Rα) that associates with the 120-140 kDa β subunit (CD131/Common β-chain/βc) to form the functional IL-3 receptor complex. The βc chain is also shared with distinct α chain subunits to form the functional heterodimeric receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on a subset of peripheral blood dendritic cells, myeloid precursors, basophils, mast cells, macrophages, and megakaryocytes. Reports indicate that IL-3Rα is also expressed on lymphocytes. The IL-3R plays an important role in hematopoietic progenitor cell growth and differentiation. This antibody does not block binding of IL-3 to the IL-3 receptor.
Spezifikation
| CD123 | |
| Monoclonal | |
| 0.2mg/mL | |
| IL-3 Receptor α chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 9F5 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IL-3Rα Transfected Cell Line | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.