Learn More
CD274 Mouse anti-Human, Brilliant Violet 786, Clone: MIH1, BD
Beschreibung
The MIH1 monoclonal antibody specifically binds to CD274, also known as Programmed Death Ligand 1 (PD-L1) and B7 Homolog 1 (B7-H1). CD274/PD-L1 and CD273/PD-L2 are members of the B7 family within the Immunoglobulin superfamily. These type I transmembrane glycoproteins function as ligands for CD279, likewise known as the Programmed Death 1 (PD-1) receptor. CD274 is expressed on activated dendritic cells, macrophages and T cells. Studies show some overlapping functions of CD274 and CD273 and indicate an important role for CD279 and its ligands in regulating T and B cell responses. CD274 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, keratinocytes, and carcinomas. The MIH1 antibody is reportedly a blocking antibody.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Spezifikation
Spezifikation
| Antigen | CD274 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | MIH1 |
| Konjugat | Brilliant Violet 786 |
| Beschreibung | B7-H1; Programmed death ligand 1; PD-L1; PDCD1 ligand 1; PDCD1L1; PDCD1LG1 |
| Zusammensetzung | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Wirtsspezies | Mouse |
| Immunogen | Human B7-H1 Transfected Cell Line |
| Reinigungsverfahren | Affinity Purified |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.