Learn More
CD45 Rat anti-Mouse, Unlabeled, Clone: 30-F11, BD
Beschreibung
The 30-F11 antibody reacts with both alloantigens and all isoforms of the CD45 leukocytes common antigen (LCA), also known as Ly-5 or T200, found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
Spezifikation
| Antigen | CD45 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 30-F11 |
| Konzentration | 0.5mg/mL |
| Konjugat | Unconjugated |
| Beschreibung | Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4 |
| Zusammensetzung | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Wirtsspezies | Rat |
| Immunogen | Mouse Thymus or Spleen |
| Mehr anzeigen |
For Research Use Only.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.