missing translation for 'onlineSavingsMsg'
Learn More

CD45 Rat anti-Mouse, FITC, Clone: 30-F11, BD

Artikelnummer. p-3969753
Change view
Click to view available options
Menge:
0.1 mg
0.5 mg
Packungsgröße:
1 Stück
Artikelnummer. Menge unitSize
10713245 0.1 mg 1 Stück
15896978 0.5 mg 1 Stück
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10713245

Marke: BD Biosciences 553079

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

The 30-F11 clone has been reported to react with all isoforms and both alloantigens of CD45, which is found on hematopoietic stem cells and all cells of hematopoietic origin, except erythrocytes. CD45 is a transmembrane glycoprotein which is expressed at high levels on the cell-surface, and its presence distinguishes leukocytes from non-hematopoietic cells. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family, where the intracellular carboxy-terminal region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated as A, B, and C, respectively). CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction and the CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 30-F11
Isotype: IgG2b κ
Species Reactivity: Mouse
Immunogen: Mouse Thymus or Spleen

Flow Cytometry

TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CD45
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon 30-F11
Konzentration 0.5mg/mL
Konjugat FITC
Beschreibung Ptprc; CD45R; LCA; Leukocyte common antigen; T200; B220; Ly-5; Lyt-4
Zusammensetzung Aqueous buffered solution containing ≤0.09% sodium azide.
Wirtsspezies Rat
Immunogen Mouse Thymus or Spleen
Reinigungsverfahren Affinity Purified
Menge 0.1 mg
Regulatorischer Status RUO
Forschungsgebiet Immunology
Primär oder sekundär Primary
Zielspezies Mouse
Inhalt und Lagerung Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Mehr anzeigen Weniger anzeigen

For Research Use Only.

Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.