missing translation for 'onlineSavingsMsg'
Learn More

CD62L Mouse anti-Human, BB515, Clone: SK11, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 565038

 View more versions of this product

Product Code. 15854039

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

The SK11 monoclonal antibody specifically binds to CD62L which is also known as, L-selectin, Leukocyte-endothelial cell adhesion molecule 1 (LECAM1/LECAM-1), Leukocyte adhesion molecule 1 (LAM1/LAM-1), Lymph node homing receptor (LNHR), Leu-8, or TQ1. CD62L is an ∽80 kDa type I transmembrane glycoprotein that belongs to the Selectin/LECAM family. CD62L is differentially expressed on T cells, B cells, monocytes, granulocytes, and a subset of NK cells. The CD62L molecule is the human homolog of the mouse lymph node homing receptor, MEL-14. CD62L plays a role in leucocyte binding to inflamed endothelium and extravasation, as well as mediating lymphocyte homing into peripheral lymphoid tissues through high endothelial postcapillary venules. Soluble CD62L can result from the proteolytic cleavage of cell surface CD62L during cellular activation or inflammation.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD62L
Monoclonal
Brilliant Blue 515 (BB515)
Aqueous buffered solution containing ≤0.09% sodium azide
Human Peripheral Blood T Lymphocytes
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
SK11
L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14
Mouse
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.