Learn More
CD62L Mouse anti-Human, BB515, Clone: SK11, BD
Mouse Monoclonal Antibody
285.00 € - 298.53 €
Spezifikation
| Antigen | CD62L |
|---|---|
| Klon | SK11 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Konjugat | Brilliant Blue 515 (BB515) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15854039
|
BD Biosciences
565038 |
25 Tests | |||||||
|
15844039
|
BD Biosciences
565037 |
100 Tests |
300.00 € 298.53 € / 1 Stück Sparen 1.47 € 0.49% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The SK11 monoclonal antibody specifically binds to CD62L which is also known as, L-selectin, Leukocyte-endothelial cell adhesion molecule 1 (LECAM1/LECAM-1), Leukocyte adhesion molecule 1 (LAM1/LAM-1), Lymph node homing receptor (LNHR), Leu-8, or TQ1. CD62L is an ∽80 kDa type I transmembrane glycoprotein that belongs to the Selectin/LECAM family. CD62L is differentially expressed on T cells, B cells, monocytes, granulocytes, and a subset of NK cells. The CD62L molecule is the human homolog of the mouse lymph node homing receptor, MEL-14. CD62L plays a role in leucocyte binding to inflamed endothelium and extravasation, as well as mediating lymphocyte homing into peripheral lymphoid tissues through high endothelial postcapillary venules. Soluble CD62L can result from the proteolytic cleavage of cell surface CD62L during cellular activation or inflammation.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Spezifikation
| CD62L | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| IgG2a κ | |
| Affinity Purified | |
| L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14 |
| SK11 | |
| Monoclonal | |
| Mouse | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Human Peripheral Blood T Lymphocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.