Learn More
CD74 molecule, major histocompatibility complex, class II invariant chain, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CD74 protein.
Marke: Abnova H00000972-D01P.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sequence: MHRRRSRSCREDQKPVMDDQRDLISNNEQLPMLGRRPGAPESKCSRGALYTGFSILVTLLLAGQATTAYFLYQQQGRLDKLTVTSQNLQLENLRMKLPKPPKPVSKMRMATPLLMQALPMGALPQGPMQNATKYGNMTEDHVMHLLQSHWNWRTRLLGWVSpezifikation
| CD74 molecule, major histocompatibility complex, class II invariant chain | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CD74 protein. | |
| CD74 | |
| DHLAG/HLADG/Ia-GAMMA | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001025158.1 | |
| CD74 | |
| CD74 (NP_001020329.1, 1 a.a. ∼ 160 a.a) full-length human protein. | |
| 100 μg | |
| Membrane Receptors | |
| 972 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |