missing translation for 'onlineSavingsMsg'
Learn More

Ceruloplasmin Mouse, Unlabeled, Clone: 8, BD

Artikelnummer. p-3970433
Ansicht ändern
Click to view available options
Menge:
50 μg
Packungsgröße:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15875949 50 μg 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15875949 Lieferant BD Biosciences Lieferanten-Nr. 611488

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Ceruloplasmin (CP) is a copper-containing glycoprotein that carries approximately 90% of plasma copper. CP contains six copper atom binding sites and is prone to transfer its copper atoms to tissues, however it is not essential for copper transport. In cultured endothelial cells, CP release of copper in the cytosol inhibits nitric oxide synthase activation, and this may be important for regulating vascular tone in blood vessels. In addition to its copper binding role, CP has also been shown to have ferroxidase activity, which involves conversion of ferrous to
ferric iron. The iron metabolism disorder aceruloplasminemia results from mutations in CP and is characterized by massive iron deposits in liver, pancreas, brain, retina, and other tissues. CP is expressed in the liver, the splenic reticuloendothelial system, the bronchiolar epithelium of the lung, and the retina and brain. The cell-specific expression of CP in neural tissues may account for the neuropathologies associated with aceruloplaminemia, while the wide expression pattern of CP may demonstrate its importance in the regulation of copper and iron levels in many tissues.

Host Species: Mouse
Clone: 8
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse ceruloplasmin aa. 233-355
Formula Weight [Chemical]: 132kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Ceruloplasmin
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 8
Konzentration 250μg/mL
Konjugat Unconjugated
Zusammensetzung Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Wirtsspezies Mouse
Immunogen Mouse ceruloplasmin aa. 233-355
Reinigungsverfahren Affinity Purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Biology
Primär oder sekundär Primary
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store undiluted at -20°C.
Isotype IgG1
Mehr anzeigen Weniger anzeigen

For Research Use Only.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.