missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA516867
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PA5-16867 targets Claudin 3 in IHC (P) and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16867 immunogen is a synthetic peptide derived from the C-terminus of mouse Claudin-3.
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
Spezifikation
| Claudin 3 | |
| Polyclonal | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| O15551, Q9Z0G9, Q63400 | |
| CLDN3 | |
| A synthetic peptide derived from the C-terminus of mouse Claudin-3 | |
| 500 μL | |
| Primary | |
| Chicken, Human, Mouse, Porcine, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CLDN3 | |
| CPE-R 2, CPE-receptor 2, Clostridium perfringens enterotoxin receptor 2, claudin-3, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12739, 1365, 65130 | |
| -20°C or -80°C if preferred | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur