Learn More
CREB1, Mouse, Clone: 2B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CREB1.
Marke: Abnova H00001385-M08.100ug
Beschreibung
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: DAAVTEAENQQMTVQAQPQIATLAQVSMPAAHATSSAPTVTLVQLPNGQTVQVHGVIQAAQPSVIQSPQVQTVQSSCKDLKRLFSGTQSpezifikation
| CREB1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC010636 | |
| CREB1 | |
| CREB1 (AAH10636.1, 14 a.a. ∼ 101 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2B2 | |
| Mouse monoclonal antibody raised against a partial recombinant CREB1. | |
| CREB1 | |
| CREB/MGC9284 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1385 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.