Learn More
cytochrome P450, family 3, subfamily A, polypeptide 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Beschreibung
This gene, CYP3A4, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. This enzyme is involved in the metabolism of approximately half the drugs which are are used today, including acetaminophen, codeine, cyclosporin A, diazepam and erythromycin. The enzyme also metabolizes some steroids and carcinogens. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Previously another CYP3A gene, CYP3A3, was thought to exist; however, it is now thought that this sequence represents a transcript variant of CYP3A4. [provided by RefSeq
Spezifikation
Spezifikation
| Antigen | cytochrome P450, family 3, subfamily A, polypeptide 4 |
| Anwendungen | Immunohistochemistry (PFA fixed), Western Blot |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Beschreibung | Mouse polyclonal antibody raised against a full-length human CYP3A4 protein. |
| Zusammensetzung | PBS with no preservative; pH 7.4 |
| Gen | CYP3A4 |
| Gen-Zugriffsnummer | NM_017460.3 |
| Gen-Alias | CP33/CP34/CYP3A/CYP3A3/HLP/MGC126680/NF-25/P450C3/P450PCN1 |
| Gensymbole | CYP3A4 |
| Mehr anzeigen |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.