Learn More
EMP3, Mouse, Clone: 3D4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant EMP3.
Marke: Abnova H00002014-M01.100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Sequence: MSLLLLVVSALHILILILLFVATLDKSWWTLPGKESLNLWYDCTWNNDTKTWACSNVSENGWLKAVQVLMVLSLILCCLSFILFMFQLYTMRRGGLFYATGLCQLCTSVAVFTGALIYAIHAEEILEKHPRGGSFGYCFALAWVAFPLALVSGIIYIHLRKRESpezifikation
| EMP3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC009718 | |
| EMP3 | |
| EMP3 (AAH09718, 1 a.a. ∼ 163 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 3D4 | |
| Mouse monoclonal antibody raised against a full length recombinant EMP3. | |
| EMP3 | |
| YMP | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2014 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |