missing translation for 'onlineSavingsMsg'
Learn More

eNOS (pS1177), Phospho-Specific Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

216.14 € - 496.48 €

Spezifikation

Antigen eNOS (pS1177), Phospho-Specific
Klon 19
Konzentration 250μg/mL
Anwendungen Western Blot
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15856189
Unterlagen ansehen
BD Biosciences
612392
50 μg
ONLINE EXCLUSIVE PRICE
218.00 €
216.14 € / 1 Stück
Sparen 1.86 €
0.85% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15866189
Unterlagen ansehen
BD Biosciences
612393
150 μg
ONLINE EXCLUSIVE PRICE
498.00 €
496.48 € / 1 Stück
Sparen 1.52 €
0.31% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits signals involved in vasorelaxation, neurotransmission, and cytotoxity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+; levels and enhance calmodulin binding. Neuronal NOS (nNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin. eNOS has a unique N-myristylation consensus sequence that may explain its membrane localization. Various protein kinases have been implicated in regulation of eNOS activity, including AMPK, PKA, PKB/Akt, PKC, and CaM Kinase II. During VEGF stimulation, eNOS is transiently phosphorylated at Ser-1177 by PKB/akt and dephosphorylated at Thr-495. At later time points, VEGF stimulation leads to an increase in Thr-495 phosphorylation mediated by PKC and a decrease in Ser-1177 phosphorylation. In addition, Ser-633 and Ser-1177 are phosphorylated by PKA and PKG in vitro. Thus, eNOS activity may be regulated through complex phosphorylated events mediated by multiple kinases at various phosphorylation sites. Human endothelial cells are routinely tested as a positive control for eNOS (pS1177) mAb. 100% homology is detected for immunogen sequence in human, mouse, rat, dog and bovine. Cross-reactivity with other species is expected but not confirmed.

Flow Cytometry, Western Blotting

Spezifikation

Spezifikation

eNOS (pS1177), Phospho-Specific
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
19
Western Blot
Unconjugated
RUO
Human
Human eNOS (pS1177)
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt