Learn More
anti-Eosinophil Major Basic Protein, Clone: BMK-13 MA135360, Invitrogen™
Alle ansehen Thermo Scientific ProdukteBeschreibung
Specifically detect Eosinophil Major Basic Protein in Human samples using Thermo Scientifc™ Pierce™ anti-Eosinophil Major Basic Protein Clone: BMK-13, validated for ELISA and IHC
The protein encoded by this gene is the predominant constituent of the crystalline core of the eosinophil granule. High levels of the proform of this protein are also present in placenta and pregnancy serum, where it exists as a complex with several other proteins including pregnancy-associated plasma protein A (PAPPA), angiotensinogen (AGT), and C3dg. This protein may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin, and in immune hypersensitivity reactions. It is directly implicated in epithelial cell damage, exfoliation, and bronchospasm in allergic diseases.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: BMK-13
Immunogen: MBP.
Formulation: 1mL of purified antibody (0.1mg/mL) in PBS buffer, with BSA 0.1%;
Uniprot Accession: P13727
Gene Symbol: PRG2
MA1-35360 detects Eosinophil Major Basic Protein in Human samples. MA1-35360 has been successfully used in ELISA and IHC procedures.MA1-35360 The immunogen is MBP.
Spezifikation
Spezifikation
| Antigen | Eosinophil Major Basic Protein |
| Anwendungen | ELISA, Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | BMK-13 |
| Konjugat | Unconjugated |
| Gensymbole | PRG2 |
| Wirtsspezies | Mouse |
| Immunogen | MBP. |
| Menge | 1 mL |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.