missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FEN1 Monoclonal Antibody (4E7)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA123228
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: MCF7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
Spezifikation
| FEN1 | |
| Monoclonal | |
| 1.73 mg/mL | |
| PBS with no preservative | |
| P39748, P39749 | |
| FEN1 | |
| Recombinant human FEN-1 protein encoding amino acids 1-380 purified E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 4E7 | |
| Unconjugated | |
| FEN1 | |
| AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 14156, 2237 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur