Learn More
Flk-1 Rat anti-Mouse, APC-Cy7, Clone: Avas 12alpha1, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561252
Beschreibung
The Avas 12 α 1 monoclonal antibody specifically binds to fetal liver kinase 1 (Flk-1) which is also known as CD309. Flk-1 is a receptor protein tyrosine kinase which is closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2 or VEGFR2), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic and adult tissues. In vivo and in vitro studies indicate that Flk-1 is required for the embryonic development of vascular endothelial and hematopoietic cells.
Spezifikation
| Flk-1 | |
| Monoclonal | |
| 0.2mg/mL | |
| CD309; Fetal liver kinase 1; Kdr; VEGF Receptor-2; VEGFR-2; AVAS12; | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| Avas 12alpha1 | |
| APC-Cyanine7 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Mouse Flk-1 Recombinant Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.